Assay Details
| Gene Symbol: MCCC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031565.1 | NP_001026736.1 | 9 - 10 | 1517 | 67 | |
| XM_025152899.1 | XP_025008667.1 | 9 - 10 | 1520 | 67 | ||
| GenBank mRNA | AJ719309.1 | - | 9 - 10 | 1517 | 67 | |
More Information
Set Membership: |
|

MCCC2L
Genomic Map