Assay Details
| Gene Symbol: CECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030767.2 | NP_001025938.1 | 2 - 3 | 433 | 68 | |
| XM_015285830.2 | XP_015141316.1 | 2 - 3 | 990 | 68 | ||
| XM_015285839.2 | XP_015141325.1 | 2 - 3 | 447 | 68 | ||
| GenBank mRNA | AJ720326.1 | - | 2 - 3 | 434 | 68 | |
| AY902779.1 | - | 2 - 3 | 436 | 68 | ||
More Information
Set Membership: |
|

CECR1
Genomic Map