Assay Details
| Gene Symbol: CHRDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204171.1 | NP_989502.1 | 5 - 6 | 866 | 70 | |
| XM_015278152.2 | XP_015133638.1 | 5 - 6 | 608 | 70 | ||
| XM_015278154.2 | XP_015133640.1 | 5 - 6 | 572 | 70 | ||
| GenBank mRNA | AF257352.1 | - | 5 - 6 | 866 | 70 | |
More Information
Set Membership: |
|

CHRDL1
Genomic Map