Assay Details
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006211.2 | NP_001006211.1 | 2 - 3 | 290 | 69 | |
| GenBank mRNA | AJ720650.1 | - | 2 - 3 | 290 | 69 | |
More Information
Set Membership: |
|

SBDS
Genomic Map