Assay Details
| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204919.1 | NP_990250.1 | 1 | 254 | 163 | |
| XM_025144276.1 | XP_025000044.1 | 1 | 289 | 163 | ||
| XM_025144284.1 | XP_025000052.1 | 1 | 289 | 163 | ||
| GenBank mRNA | AF038560.1 | - | 1 | 254 | 163 | |
More Information
Set Membership: |
|

MAP6
Genomic Map