Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031413.1 | NP_001026584.1 | 2 | 255 | 103 | |
| GenBank mRNA | AJ720741.1 | - | 2 | 255 | 103 | |
| CR386877.1 | - | 2 | 280 | 103 | ||
More Information
Set Membership: |
|

DOHH
Genomic Map