Assay Details
| Gene Symbol: NUDT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252066.2 | NP_001238995.2 | 2 - 3 | 221 | 74 | |
| XM_015292509.2 | XP_015147995.2 | 1 - 2 | 507 | 74 | ||
| GenBank mRNA | AM071237.1 | - | 2 - 3 | 191 | 74 | |
More Information
Set Membership: |
|

NUDT7
Genomic Map