Assay Details
| Gene Symbol: TUBA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305140.1 | NP_001292069.1 | 511 | 101 | ||
| GenBank mRNA | DR416742.1 | - | 290 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBA5| Gene Symbol: TUBA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305140.1 | NP_001292069.1 | 511 | 101 | ||
| GenBank mRNA | DR416742.1 | - | 290 | 101 | ||
Set Membership: |
|