Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278655.2 | XP_015134141.2 | 4 - 5 | 327 | 65 | |
| XM_025150334.1 | XP_025006102.1 | 4 - 5 | 1235 | 65 | ||
| XM_025150335.1 | XP_025006103.1 | 5 - 6 | 1027 | 65 | ||
| GenBank mRNA | BX929630.1 | - | 5 - 6 | 929 | 65 | |
| CR352912.1 | - | 3 - 4 | 234 | 65 | ||
More Information
Set Membership: |
|

RBMX2
Genomic Map