Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940895.3 | XP_004940952.1 | 3 - 4 | 330 | 98 | |
| XM_420372.6 | XP_420372.3 | 3 - 4 | 340 | 98 | ||
| GenBank mRNA | CR353731.1 | - | 2 - 3 | 540 | 98 | |
More Information
Set Membership: |
|

RBM46
Genomic Map