Assay Details
| Gene Symbol: TOE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422434.6 | XP_422434.2 | 6 - 7 | 853 | 97 | |
| GenBank mRNA | CR354328.1 | - | 1 - 2 | 1384 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOE1| Gene Symbol: TOE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422434.6 | XP_422434.2 | 6 - 7 | 853 | 97 | |
| GenBank mRNA | CR354328.1 | - | 1 - 2 | 1384 | 97 | |
Set Membership: |
|