Assay Details
| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142796.1 | XP_024998564.1 | 5 - 6 | 445 | 60 | |
| GenBank mRNA | AJ721038.1 | - | 5 - 6 | 413 | 60 | |
More Information
Set Membership: |
|

PBDC1
Genomic Map