Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642702.4 | XP_003642750.1 | 5 - 6 | 776 | 65 | |
| XM_003642703.4 | XP_003642751.1 | 6 - 7 | 802 | 65 | ||
| XM_003642704.4 | XP_003642752.1 | 6 - 7 | 797 | 65 | ||
| XM_003642705.4 | XP_003642753.1 | 5 - 6 | 685 | 65 | ||
| XM_004948398.2 | XP_004948455.1 | 6 - 7 | 708 | 65 | ||
| XM_015280016.2 | XP_015135502.1 | 5 - 6 | 680 | 65 | ||
| XM_025143436.1 | XP_024999204.1 | 6 - 7 | 789 | 65 | ||
| XM_025143437.1 | XP_024999205.1 | 6 - 7 | 854 | 65 | ||
| XM_427122.6 | XP_427122.2 | 5 - 6 | 708 | 65 | ||
| GenBank mRNA | BX933097.1 | - | 6 - 7 | 751 | 65 | |
| BX934755.2 | - | 6 - 7 | 760 | 65 | ||
| BX935158.1 | - | 5 - 6 | 695 | 65 | ||
| BX936145.1 | - | 6 - 7 | 745 | 65 | ||
| CR389606.1 | - | 5 - 6 | 687 | 65 | ||
More Information
Set Membership: |
|

NIT1
Genomic Map