Assay Details
| Gene Symbol: CHST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292473.2 | XP_015147959.1 | 1 - 2 | 101 | 69 | |
| GenBank mRNA | BX933194.1 | - | 1 - 2 | 60 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015292473.2 transcripts.
Set Membership: |
|

CHST8
Genomic Map