Assay Details
| Gene Symbol: FAM19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234078.5 | XP_001234079.2 | 5 - 6 | 1034 | 100 | |
| XM_004944676.3 | XP_004944733.2 | 4 - 5 | 952 | 100 | ||
| XM_015293255.2 | XP_015148741.2 | 5 - 6 | 652 | 100 | ||
| XM_015293256.2 | XP_015148742.1 | 4 - 5 | 408 | 100 | ||
| XM_015293257.2 | XP_015148743.2 | 4 - 5 | 555 | 100 | ||
| XM_015293258.2 | XP_015148744.1 | 6 - 7 | 560 | 100 | ||
| GenBank mRNA | BX936093.2 | - | 4 - 5 | 403 | 100 | |
| CR353412.1 | - | 5 - 6 | 835 | 100 | ||
More Information
Set Membership: |
|

FAM19A1
Genomic Map