Assay Details
| Gene Symbol: ATP5J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX931387.1 | - | 1 | 220 | 87 | |
| BX931482.2 | - | 1 | 206 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5J| Gene Symbol: ATP5J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX931387.1 | - | 1 | 220 | 87 | |
| BX931482.2 | - | 1 | 206 | 87 | ||
Set Membership: |
|