Assay Details
| Gene Symbol: HEMG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249447.1 | NP_001236376.1 | 16 - 17 | 1313 | 95 | |
| GenBank mRNA | AB025102.1 | - | 16 - 17 | 1313 | 95 | |
More Information
Set Membership: |
|

HEMG
Genomic Map