Assay Details
| Gene Symbol: PDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249840.1 | NP_001236769.1 | 8 - 9 | 1430 | 76 | |
| GenBank mRNA | M64704.1 | - | 8 - 9 | 1430 | 76 | |
More Information
Set Membership: |
|

PDS1
Genomic Map