Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249421.1 | NP_001236350.1 | 3 - 4 | 849 | 101 | |
| GenBank mRNA | AJ871579.1 | - | 3 - 4 | 849 | 101 | |
| AK245180.1 | - | 2 - 3 | 277 | 101 | ||
| AK245673.1 | - | 3 - 4 | 862 | 101 | ||
| Gene Symbol: LOC100818063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245644.1 | - | 3 - 4 | 882 | 101 | |
More Information
Set Membership: |
|

BMY2
Genomic Map