Assay Details
| Gene Symbol: LOC100803259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289334.1 | NP_001276263.1 | 1 | 145 | 71 | |
| GenBank mRNA | AK285965.1 | - | 1 | 216 | 71 | |
| BT094608.1 | - | 1 | 147 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT094608.1, NM_001289334.1 transcripts.
Set Membership: |
|

LOC100803259
Genomic Map