Assay Details
| Gene Symbol: Kif2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872899.2 | XP_004872956.1 | 8 - 9 | 867 | 73 | |
| XM_004872901.3 | XP_004872958.1 | 7 - 8 | 895 | 73 | ||
| XM_021250777.1 | XP_021106436.1 | 8 - 9 | 868 | 73 | ||
| XM_021250778.1 | XP_021106437.1 | 7 - 8 | 609 | 73 | ||
More Information
Set Membership: |
|

Kif2c
Genomic Map