Assay Details
| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872915.3 | XP_004872972.1 | 9 - 10 | 1116 | 60 | |
| XM_021250807.1 | XP_021106466.1 | 7 - 8 | 904 | 60 | ||
| XM_021250808.1 | XP_021106467.1 | 9 - 10 | 1230 | 60 | ||
More Information
Set Membership: |
|

Urod
Genomic Map