Assay Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858459.3 | XP_004858516.1 | 4 - 5 | 1172 | 62 | |
| XM_013077227.2 | XP_012932681.1 | 4 - 5 | 1170 | 62 | ||
| XM_021239263.1 | XP_021094922.1 | 4 - 5 | 1173 | 62 | ||
More Information
Set Membership: |
|

Cdh19
Genomic Map