Assay Details
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853529.3 | XP_004853586.2 | 5 - 6 | 866 | 67 | |
| XM_004853530.3 | XP_004853587.2 | 4 - 5 | 753 | 67 | ||
| XM_004853531.3 | XP_004853588.2 | 4 - 5 | 753 | 67 | ||
| XM_021265552.1 | XP_021121211.1 | 5 - 6 | 854 | 67 | ||
More Information
Set Membership: |
|

Lpgat1
Genomic Map