Assay Details
| Gene Symbol: Nup62cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241578.1 | XP_021097237.1 | 4 - 5 | 730 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup62cl| Gene Symbol: Nup62cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241578.1 | XP_021097237.1 | 4 - 5 | 730 | 103 | |
Set Membership: |
|