Assay Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836471.2 | XP_004836528.1 | 2 - 3 | 359 | 78 | |
| XM_013078651.2 | XP_012934105.1 | 2 - 3 | 484 | 78 | ||
| XM_021240397.1 | XP_021096056.1 | 2 - 3 | 362 | 78 | ||
| XM_021240406.1 | XP_021096065.1 | 2 - 3 | 361 | 78 | ||
More Information
Set Membership: |
|

Ublcp1
Genomic Map