Assay Details
| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850951.3 | XP_004851008.1 | 3 - 4 | 453 | 76 | |
| XM_021263322.1 | XP_021118981.1 | 4 - 5 | 516 | 76 | ||
| XM_021263323.1 | XP_021118982.1 | 3 - 4 | 289 | 76 | ||
More Information
Set Membership: |
|

Trappc3
Genomic Map