Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845329.2 | XP_004845386.1 | 9 - 10 | 874 | 66 | |
| XM_004845330.2 | XP_004845387.1 | 9 - 10 | 872 | 66 | ||
| XM_004845331.2 | XP_004845388.1 | 8 - 9 | 828 | 66 | ||
More Information
Set Membership: |
|

Mapk12
Genomic Map