Assay Details
| Gene Symbol: Map3k21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854637.3 | XP_004854694.1 | 5 - 6 | 1737 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k21| Gene Symbol: Map3k21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854637.3 | XP_004854694.1 | 5 - 6 | 1737 | 63 | |
Set Membership: |
|