Assay Details
| Gene Symbol: LOC101726503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847528.3 | XP_004847585.1 | 1 - 2 | 1191 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726503| Gene Symbol: LOC101726503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847528.3 | XP_004847585.1 | 1 - 2 | 1191 | 106 | |
Set Membership: |
|