Assay Details
| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858747.2 | XP_004858804.1 | 6 - 7 | 7899 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vsig8| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858747.2 | XP_004858804.1 | 6 - 7 | 7899 | 63 | |
Set Membership: |
|