Assay Details
| Gene Symbol: Wdr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252695.1 | XP_021108354.1 | 5 - 6 | 1713 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr97| Gene Symbol: Wdr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252695.1 | XP_021108354.1 | 5 - 6 | 1713 | 56 | |
Set Membership: |
|