Assay Details
| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839560.3 | XP_004839617.1 | 9 - 10 | 1567 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpnmb| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839560.3 | XP_004839617.1 | 9 - 10 | 1567 | 60 | |
Set Membership: |
|