Assay Details
| Gene Symbol: Polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874583.2 | XP_004874640.1 | 1 - 2 | 199 | 60 | |
| XM_013069176.2 | XP_012924630.1 | 1 - 2 | 199 | 60 | ||
| XM_021251994.1 | XP_021107653.1 | 1 - 2 | 199 | 60 | ||
More Information
Set Membership: |
|

Polr1e
Genomic Map