Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239805.1 | XP_021095464.1 | 13 - 14 | 2563 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd22| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239805.1 | XP_021095464.1 | 13 - 14 | 2563 | 61 | |
Set Membership: |
|