Assay Details
| Gene Symbol: Erich6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257774.1 | XP_021113433.1 | 9 - 10 | 1511 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich6b| Gene Symbol: Erich6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257774.1 | XP_021113433.1 | 9 - 10 | 1511 | 87 | |