Assay Details
| Gene Symbol: Fam19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854175.3 | XP_004854232.1 | 3 - 4 | 435 | 73 | |
| XM_004854176.3 | XP_004854233.1 | 3 - 4 | 432 | 73 | ||
| XM_021266017.1 | XP_021121676.1 | 3 - 4 | 327 | 73 | ||
More Information
Set Membership: |
|

Fam19a1
Genomic Map