Assay Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850334.3 | XP_004850391.1 | 5 - 6 | 1089 | 57 | |
| XM_004850335.3 | XP_004850392.1 | 5 - 6 | 1091 | 57 | ||
More Information
Set Membership: |
|

Agmat
Genomic Map