Assay Details
| Gene Symbol: Dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838209.2 | XP_004838266.1 | 1 - 2 | 752 | 74 | |
| XM_021252750.1 | XP_021108409.1 | 1 - 2 | 749 | 74 | ||
More Information
Set Membership: |
|

Dmrt1
Genomic Map