| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000485588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000485588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000621603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
ENST00000621603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
TC14002309.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
ENST00000393078.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
ENST00000393080.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
ENST00000467132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
ENST00000482740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007313.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000485588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000485588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000555820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000621603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
ENST00000621603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010644.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000393078.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000393080.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000467132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000482740.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000556388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
ENST00000556968.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
NM_001085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400180401.hg.1 |
TC1400010644.hg.1 |
Clariom D TC |