| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003884.hg.1 |
TC0X000290.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003884.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003886.hg.1 |
TC0X000290.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003886.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003886.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003885.hg.1 |
TC0X000290.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003885.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003885.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007256.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149958.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149958.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149958.hg.1 |
TC0X00007256.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00149959.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149959.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149959.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149959.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149959.hg.1 |
TC0X00007256.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00149960.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149960.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149960.hg.1 |
ENST00000252677.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149960.hg.1 |
NM_005448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149960.hg.1 |
TC0X00007256.hg.1 |
Clariom D TC |