Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031304.4 | NP_112594.1 | 1 - 2 | 95 | 100 | |
| GenBank mRNA | BC002817.2 | - | 1 - 2 | 95 | 3635058 | 100 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001043.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC002817.2,NM_031304.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014218.hg.1 | ENST00000250937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014218.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014218.hg.1 | TC19001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014219.hg.1 | ENST00000250937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014219.hg.1 | ENST00000427575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014219.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014219.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014219.hg.1 | TC19001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014216.hg.1 | ENST00000250937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014216.hg.1 | ENST00000427575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014216.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014216.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014216.hg.1 | TC19001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174024.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174023.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174023.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174023.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174023.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174023.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174021.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174020.hg.1 | NM_031304.4 | Refseq |

DOHH
Genomic Map