Assay Details
| Gene Symbol: TTTY5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001541.1 | 1 - 2 | 576 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0Y000194.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 540413 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 117250 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34209 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P34209 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 4175 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36383 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34209 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P34209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15002318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12067203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32847787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17466518 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P34209 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P34209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067211 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466446 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466502 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17482985 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067207 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466444 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17472328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067204 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466520 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067200 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466448 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067183 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466470 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466473 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466475 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466482 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17468833 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067198 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466455 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12067188 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17466530 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15002322 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P17403159 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_001541_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8177345 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8177346 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8177344 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224040_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11745588_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11745588_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006764.hg.1 | ENST00000558356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006764.hg.1 | ENST00000558356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001986.hg.1 | TC0Y000194.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001986.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001986.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007205.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007205.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007205.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007205.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001987.hg.1 | TC0Y000194.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001987.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y001987.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00007205.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007205.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007205.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00007205.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006764.hg.1 | ENST00000558356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006764.hg.1 | ENST00000558356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00148458.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00148458.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00148458.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00148458.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00148458.hg.1 | TC0Y00007205.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0Y00148457.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00148457.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00148457.hg.1 | ENST00000400581.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00148457.hg.1 | NR_001541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00148457.hg.1 | TC0Y00007205.hg.1 | Clariom D TC |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_001541_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041940 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 165150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1685986 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1685986 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7610500 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_8607 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2164478 | NR_001541.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2164482 | NR_001541.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1685986 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2164478 | NR_001541.1 | human_transcriptome |

TTTY5
Genomic Map