| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000307.hg.1 |
TC07000023.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000307.hg.1 |
ENST00000343242.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000307.hg.1 |
ENST00000403150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000307.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000304.hg.1 |
TC07000023.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000304.hg.1 |
ENST00000343242.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000304.hg.1 |
ENST00000403150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000304.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000305.hg.1 |
TC07000023.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000305.hg.1 |
ENST00000343242.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000305.hg.1 |
ENST00000403150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000305.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144686.hg.1 |
TC0700006520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144689.hg.1 |
TC0700006520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144688.hg.1 |
TC0700006520.hg.1 |
Clariom D TC |