Assay Details
| Gene Symbol: LOC150005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057475.1 | - | 1 | 255 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287928 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287910 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946227 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287936 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287911 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946249 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287933 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17222517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946226 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11287932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31946221 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3482461 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44154 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3482461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1561342_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1561343_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561342_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561342_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561343_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561343_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561343_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561342_at7 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1877412 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5550349 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_75089 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1824864 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1824864 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1877412 | - | - |

LOC150005
Genomic Map