Assay Details
| Gene Symbol: STARD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525821.1 | XP_011524123.1 | 1 - 2 | 98 | 141 | |
| XM_017025552.1 | XP_016881041.1 | 1 - 2 | 98 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525821.1, XM_017025552.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |

STARD6
Genomic Map