Assay Details
| Gene Symbol: ZCCHC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185059.1 | NP_001171988.1 | 6 - 7 | 1219 | 148 | |
| NM_024617.3 | NP_078893.2 | 6 - 7 | 1329 | 148 | ||
| XM_005252207.3 | XP_005252264.1 | 6 - 7 | 1281 | 148 | ||
| XM_005252208.2 | XP_005252265.1 | 6 - 7 | 1281 | 148 | ||
| XM_006717283.1 | XP_006717346.1 | 6 - 7 | 1281 | 148 | ||
| XM_011519011.2 | XP_011517313.1 | 6 - 7 | 1281 | 148 | ||
| XM_011519012.2 | XP_011517314.1 | 6 - 7 | 1229 | 148 | ||
| XM_011519013.2 | XP_011517315.1 | 6 - 7 | 1898 | 148 | ||
| XM_011519014.2 | XP_011517316.1 | 6 - 7 | 1500 | 148 | ||
| XM_011519015.1 | XP_011517317.1 | 6 - 7 | 1281 | 148 | ||
| XM_011519016.1 | XP_011517318.1 | 6 - 7 | 1281 | 148 | ||
| XM_017015130.1 | XP_016870619.1 | 6 - 7 | 1897 | 148 | ||
| XM_017015131.1 | XP_016870620.1 | 6 - 7 | 1539 | 148 | ||
| XM_017015132.1 | XP_016870621.1 | 6 - 7 | 1281 | 148 | ||
| GenBank mRNA | AK055546.1 | - | 6 - 7 | 1254 | 148 | |
| AL832193.1 | - | 6 - 7 | 1253 | 148 | ||
| BC032456.1 | - | 6 - 7 | 1236 | 148 | ||
| BC144523.1 | - | 5 - 6 | 1120 | 148 | ||
| BX641077.1 | - | 6 - 7 | 1305 | 148 | ||
| CR933643.1 | - | 6 - 7 | 1196 | 148 | ||
| CR933644.1 | - | 6 - 7 | 1256 | 148 | ||
| CR936608.1 | - | 6 - 7 | 1253 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017015132.1 transcripts.
Set Membership: |
|

ZCCHC6
Genomic Map