Assay Details
| Gene Symbol: HLA-DRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022555.3 | NP_072049.2 | 5 - 6 | 827 | 148 | |
| GenBank mRNA | A06800.1 | - | 5 - 6 | 823 | 148 | |
| AF000448.1 | - | 5 - 6 | 700 | 148 | ||
| BC008987.1 | - | 5 - 6 | 811 | 4184806 | 148 | |
| BC106057.1 | - | 5 - 6 | 890 | 4385464 | 148 | |
| M28991.1 | - | 5 - 6 | 745 | 146 | ||
| M28992.1 | - | 5 - 6 | 706 | 146 | ||
| V00522.1 | - | 5 - 6 | 822 | 148 | ||
| Gene Symbol: HLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M26038.1 | - | 5 - 6 | 833 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3383912 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P145336 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P402222 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 18862 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 26133 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P138377 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P138377 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6396 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P343233 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3383912 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3383912 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P402222 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P145336 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P402222 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P145336 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6303 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 3004 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 8169 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_022555_at | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8180003 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8178802 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003800.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003800.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003800.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003807.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003807.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003809.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003809.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003808.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003808.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003801.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003801.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003801.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003803.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003803.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_022555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_001243965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC6_cox_hap2000193.hg.1 | NM_002124.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003810.hg.1 | TC6_cox_hap2000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003810.hg.1 | NM_022555.3 | Refseq |
| Human Genome U133 Plus 2.0 Array | 215289_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 206111_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 238900_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215193_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215289_at | - | - |
| Human Genome U133 Plus 2.0 Array | 204670_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209312_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216667_at | - | - |
| Human Genome U133 Plus 2.0 Array | 206111_at | - | - |
| Human Genome U133 Plus 2.0 Array | 204670_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221491_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215193_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209312_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216667_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 238900_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221491_x_at | - | - |
| Human Genome U133A 2.0 Array | 204670_x_at | - | - |
| Human Genome U133A 2.0 Array | 216667_PM_at | - | - |
| Human Genome U133A 2.0 Array | 209312_x_at | - | - |
| Human Genome U133A 2.0 Array | 206111_PM_at | - | - |
| Human Genome U133A 2.0 Array | 215289_PM_at | - | - |
| Human Genome U133A 2.0 Array | 204670_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 216667_at | - | - |
| Human Genome U133A 2.0 Array | 215193_x_at | - | - |
| Human Genome U133A 2.0 Array | 215193_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 221491_x_at | - | - |
| Human Genome U133A 2.0 Array | 206111_at | - | - |
| Human Genome U133A 2.0 Array | 209312_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 215289_at | - | - |
| Human Genome U133A 2.0 Array | 221491_PM_x_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_022555_at | - | - |
| Human Genome U95 Set | 41723_s_at | - | - |
| Human Genome U95 Set | 33261_at | - | - |
| Human Genome U95 Set | 31336_PM_at | - | - |
| Human Genome U95 Set | 36766_at | - | - |
| Human Genome U95 Set | 41723_PM_s_at | - | - |
| Human Genome U95 Set | 31336_at | - | - |
| Human Genome U95 Set | 33261_PM_at | - | - |
| Human Genome U95 Set | 36766_PM_at | - | - |
| Human Genome U95 Set | 63416_s_at | - | - |
| Human Genome U95 Set | 54906_PM_r_at | - | - |
| Human Genome U95 Set | 54906_r_at | - | - |
| Human Genome U95 Set | 63416_PM_s_at | - | - |
| Human Genome U95 Set | 75284_f_at | - | - |
| Human Genome U95 Set | 75284_PM_f_at | - | - |
| Human Genome U95 Set | 90319_at | - | - |
| Human Genome U95 Set | 90319_PM_at | - | - |
| Human Genome U133 Set | 204670_PM_x_at | - | - |
| Human Genome U133 Set | 209312_PM_x_at | - | - |
| Human Genome U133 Set | 215289_PM_at | - | - |
| Human Genome U133 Set | 215193_x_at | - | - |
| Human Genome U133 Set | 216667_PM_at | - | - |
| Human Genome U133 Set | 204670_x_at | - | - |
| Human Genome U133 Set | 221491_x_at | - | - |
| Human Genome U133 Set | 215193_PM_x_at | - | - |
| Human Genome U133 Set | 206111_at | - | - |
| Human Genome U133 Set | 221491_PM_x_at | - | - |
| Human Genome U133 Set | 215289_at | - | - |
| Human Genome U133 Set | 216667_at | - | - |
| Human Genome U133 Set | 209312_x_at | - | - |
| Human Genome U133 Set | 206111_PM_at | - | - |
| Human Genome U133 Set | 238900_at | - | - |
| Human Genome U133 Set | 238900_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1717261 | M28991.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1717261 | M28992.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1717261 | V00522.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1717261 | A06800.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_18641371-S | M28991.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_18641371-S | M28992.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_18641371-S | V00522.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_18641371-S | A06800.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_18641371-S | CR592036.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_2942-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002340239 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002340239 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4010053 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_2942 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717261 | CR592036.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717261 | M28991.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717261 | M28992.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717261 | V00522.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717261 | A06800.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717261 | CR592036.1 | human_transcriptome |

HLA-DRB3
Genomic Map