Assay Details
| Gene Symbol: KIF26B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042481.1 | - | 3 - 4 | 892 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |

KIF26B
Genomic Map