Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130716.1 | NP_001124188.1 | 1 - 2 | 133 | 122 | |
| GenBank mRNA | BC012205.1 | - | 1 - 2 | 117 | 3859609 | 122 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001344.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC012205.1,NM_001130716.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell proliferation
response to cold
negative regulation of multicellular organism growth
defense response to bacterium
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
chromatin binding
positive regulation of cell proliferation
response to cold
negative regulation of multicellular organism growth
defense response to bacterium
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
chromatin binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018642.hg.1 | TC04001344.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018642.hg.1 | ENST00000411416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018642.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | TC04001344.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | ENST00000311507.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | ENST00000411416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | ENST00000505406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04018639.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176392.hg.1 | TC0400011192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400176393.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176393.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176393.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176393.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176393.hg.1 | TC0400011192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000509973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | ENST00000515389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011192.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000311507.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000411416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000426923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | ENST00000505406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_001130715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_001130716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | NM_016619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400176391.hg.1 | TC0400011192.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 157022 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 157102 | - | - |

PLAC8
Genomic Map